Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.24
grade
certified reference material
manufacturer/tradename
National Research Council of Canada (NRC)
application(s)
food and beverages
format
multi-component solution
General description
RM-RILC is a reference material (RM) designed for measurement of liquid chromatography (LC) retention indices (RIs) based on N-alkylpyridinium-3-sulfonates (NAPS).
Application
The N-alkylpyridinium-3-sulfonates (NAPS) mix has proven to be an effective tool to normalize liquid chromatography-based mycotoxin retention times across various laboratories. (M.W. Sumarah et al, Journal of Chromatography A Volume 1745, 29 March 2025, 465732)
Also it has been demonstrated, that the NAPS mix is effective in improving metabolite annotation and identification across different analytical conditions. (M. Witting et al Analytical and Bioanalytical Chemistry, Volume 414, pages 7387–7398, (2022))
Also it has been demonstrated, that the NAPS mix is effective in improving metabolite annotation and identification across different analytical conditions. (M. Witting et al Analytical and Bioanalytical Chemistry, Volume 414, pages 7387–7398, (2022))
Features and Benefits
NRC Canada’s certified reference materials (CRMs) are traceable to the International System of Units, and conform to the requirements of ISO/IEC 17025 and ISO 17034.For specific values, as well as additional information regarding stability and storage information, please view the Certificate of Analysis.With NRC Canada’s CRMs, you gain access to products backed by extensive research and expertise, fostering trust and consistency in your results. Elevate your laboratory’s accuracy and credibility with NRC Canada’s certified reference materials.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
For more information, please refer to the Certificate of Analysis or Safety Data Sheet
Sales restrictions may apply.
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
49.5 °F - closed cup
flash_point_c
9.7 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| RM-RILC | 04065270773395 |


