MilliporeSigma
Blood Typing
MilliporeSigma is a leading provider of monoclonal antibodies to the blood typing market. Our large portfolio of standardized antibody intermediates for further manufacturing use (FFMU) and finished reagents allows manufacturers to create and manufacture a wide variety of blood typing reagents and devices. Our brand is globally synonymous with quality backed by our ISO-certified, FDA-licensed manufacturing facilities and experienced personnel. This enables us to stay in the forefront of the industry and ensures the quality of these antibodies to meet global regulatory requirements.
Three bottles of monoclonal antibodies labeled Anti-A, Anti-B, and Anti-D, each in different colored solutions (blue, yellow, and clear) with black droppers on top. The labels include the product names and clone information.
Blood Typing Finished Reagents
We offer a large portfolio of finished reagents compliant with the requirements of the European Regulation 2017/746. These reagents are offered in a wide range of commercial options, including bulk, reagents for research use, BIOSCOT®-labelled vials carrying the CE mark or unlabeled vials.
Blood Typing Intermediates (FFMU)
Our portfolio of blood typing intermediates for further manufacturing use (FFMU) are standardized cell culture supernatants from cell lines expressing human or murine monoclonal antibodies. These FFMU intermediates are for use in formulating finished blood typing reagents or devices.
Blood Typing Intermediates for Further Manufacturing Use (FFMU)