MilliporeSigma
HomeServicesSoftware & Digital PlatformsM-Trace® Electronic Test Record Software
M-Trace® Electronic Test Record Software
The M-Trace® Software & Mobile App is your 21 CFR Part 11 compliant Digital QC companion for Sterility Testing, for trusted and efficient data traceability! Watch the videos to learn more, and see how a fully traceable, reliable and efficient sterility test performed with the assistance of the M-Trace® software could look like at your site.
Get introduced to your M-Trace® digital QC companion
Experience a sterility test using the M-Trace® solution
Filling the gap in Sterility Testing: Traceability and 21 CFR Part 11 Compliance
By giving step-by-step guidance and recording data automatically in realtime, the M-Trace® system offers a convenient and reliable way to comply with regulatory requirements regarding traceability and data integrity.
  • Connect the M-Trace® software to your current Steritest® Symbio pump(s)
  • Run the software from any computer using your centralized user management for Microsoft Windows
  • Benefit from enhanced 21 CFR part 11 compliance thanks to the audit trail function
  • Ensure full traceability and regulatory compliance from real-time, automatic, and complete recording of data for each of your sterility tests
  • Increase process safety with guided workflows and deviation alerts
  • Improve efficiency by eliminating paper-based documentation and digitizing recording and review processes
  • Facilitate automated control of your Steritest® Symbio pump(s) and greater connectivity of your testing data to your lab IT environment (one single software to control all your pumps)
  • Simplify training of operators with comprehensive real time instructions
Get full control of your entire Sterility Testing workflow
Sterility Testing workflow creation with the M-Trace software
Get your Workflow Digitalized
Digitalize your SOPs by creating fully customizable Master Test Records (MTRs) that will guide you through the workflow.
M-Trace<sup>®</sup> mobile app
Connect information on samples, consumables, test points or devices, and test results from the lab or anywhere with the M-Trace® Mobile App.
Confirm by voice control all your Sterility testing steps
Confirm the proper execution of a work step either by voice control or by machine-to-machine communication to allow real time documentation while your hands are busy with other tasks.
Obtain Steritest Symbio pump parameters to monitor your process
Obtain instrument parameters such as pump speed or pressure from instruments such as the Steritest® Symbio pump.
Generate automatically a digital test report of your sterility testing
A comprehensive report is automatically created containing all information on consumables used, incidents that occurred, and confirmation  that each work step has been carried out as defined.
21 CFR part 11 compliant user management system
Define roles and user rights in the comprehensive and convenient 21 CFR part 11 compliant user management system.
Related Products
Loading
DISCOVER M-TRACE® ELECTRONIC TEST RECORD SOFTWARE
Contact us for a demo or more information on the new M-Trace® Electronic Test Record Software.
Simply fill out and submit the form below.
Related Product Resources
Mtrace Brochure
Brochure: A deeper understanding of our M-Trace® Electronic Test Record Software
Get more detailed information and learn how a fully traceable, reliable and efficient sterility test performed with the assistance of our M-Trace® software could look like at your site. 
Download Now
Mtrace Datasheet
Datasheet: Technical information about our M-Trace® solution
Get additional technical information about your digital QC companion for sterility testing and other regulated processes.
Download Now 
Mtrace Software
Whitepaper: M-Trace® Electronic Test Record Software and regulatory compliance
Get access to the detailed analysis related to 21 CFR Part 11 compliance of our M-Trace® software.
Download Now 
Related Webinars
A person in a white lab coat holding a smartphone while observing a cart with various laboratory bottles against a blue backdrop.
Webinar: Filling the Gap in Sterility Testing: Traceability and 21 CFR Part 11 Compliance
Listen to our experts and explore how you could benefit from using our M-Trace®solution.
A person in a white lab coat manipulating a molecular model or DNA strand, with a digital background of DNA and data patterns.
Webinar: Data Integrity in the Quality Control Lab: No Pen, No Pain
Learn how innovative technologies and digitalized solutions can address data integrity challenges in microbiology Quality Control Labs.