Determination of Water Content By Karl Fischer Method in Magnesium Chloride
Product Group
Inorganic compounds
Section Overview
- General Information for Water Determination in Inorganic Compounds
- Methods for Water Determination in Magnesium Chloride Sample
- Related Materials
General Information for Water Determination in Inorganic Compounds
Salts contain water in different kinds of bonding states: adherent moisture - enclosed water - crystal water. For determination of the total water content, the substance must be completely dissolved. If the salt does not dissolve in methanol sufficiently, formamide can be added to the recipient vessel and/or titration carried out at 50 °C. If adherent moisture alone is of interest, the dissolution of the sample must be prevented through the addition of a non-polar solvent (e.g., chloroform) to the Karl Fischer solvent. A good alternative for salts to the direct volumetric determination is often the Karl Fischer oven technique. Through slow heating of the sample, it is hereby possible to differentiate among the various types of water bonding. For salts that cause disturbing side reactions in the KF solutions (e.g., carbonates, sulphites, borates) the Karl Fischer oven method is the method of choice. The same is also true for oxides and hydroxides. These alkaline compounds react with the weakly acidic KF solution, resulting in the formation of water, which is erroneously detected as water during the titration. Aqueous inorganic acids and bases are titrated volumetrically. Since stoichiometry and reaction rate of the Karl Fischer reaction are influenced by the pH, sufficient buffering of the KF solution is a prerequisite for reproducible and accurate results. Optimum conditions are obtained in the neutral pH range (pH 4 - 8). Thus, the determination of acids is performed through the addition of Aquastar® buffer solution for strong acids, imidazole or a derivative of imidazole to the titration media. For the titration of bases the pH is adjusted by addition of Aquastar® buffer solution for strong bases, salicylic acid or benzoic acid. With particularly strong, highly concentrated acids the propensity for esterification in methanolic solutions should also be noted. In such cases external neutralisation in a methanol-free medium is necessary.
Methods for Water Determination in Magnesium Chloride Sample
Magnesium chloride is only slightly soluble in common Karl Fischer reagents.
The addition of formamide as a solubilizer leads to better solubility and reproducible results.
The indirect coulometric titration in combination with the Karl Fischer oven technique shows reproducible results at 250 °C.
One-Component Volumetric Karl Fischer Titration
Karl Fischer Reagents:
- Karl Fischer Titrant
Aquastar® - CombiTitrant 5 (1.88005) - One component reagent for volumetric Karl Fischer titration, 1 mL = approx. 5 mg water
- Solvent
30 mL Aquastar® - CombiMethanol (1.88009) - Solvent for volumetric Karl Fischer titration with one component reagents, max. 0.01 % water
and
20 mL Formamide (1.09684)- as a solubilizer
Titration Parameters:
Stirring time: 180 sec.
Default titration settings, e.g.:
I(pol) = 20 - 50 µA, U(EP) = 100 - 250 mV
Stop criterion: drift < 20 µL/min
Sample Size:
30 mg
Titration Procedure:
The titration medium is first placed into the titration cell and titrated dry by means of the titrant. Then the sample is added from a weighing boat (exact sample weight determination by weighing of weighing boat before and after addition) and the titration is started. For complete extraction of the sample a stirring time of at least 180 seconds is recommended.