MilliporeSigma
HomeqPCRKiCqStart® SYBR® Green Primers FAQs
KiCqStart® SYBR® Green Primers FAQs
Compatible with any thermal cycler, KiCqStart® Primers (Product No. KSPQ12012) are ready-to-order, pre-designed primer pairs for quantifying gene expression by SYBR Green I RT-qPCR (Reverse Transcription Quantitative Real-Time PCR). To comply with MIQE, the primers have been developed using sophisticated bioinformatics tools and validated in silico to avoid off-target amplification. KiCqStart® Primers are available as up to three ranked sets of forward and reverse primer pairs for all genes from common model organisms.
What does validated in silico mean?
The performance of the sequences against the targets was verified with computational PCR. Given that the chemistry of PCR is programmed into the software, this ‘computer test tube’ is an excellent surrogate for the expected outcome of any in vitro reaction. In addition, a select number of primer pairs have undergone wet-lab validation experiments and all performed well. 
Among other things, the exon locations, sequences, and quantities of the oligonucleotides are provided either before or after purchase.
Most amplicons are between 75 and 200 base pairs.
No, the primer pairs have been designed to detect the most prevalent one for each eukaryotic gene based on a literature review. However, it is possible that the primer pairs may co-amplify other transcripts.
Search GenBank with the primer pair RefSeq.
Search GenBank with the primer pair RefSeq.
No, the primer pairs have been specifically designed for SYBR® Green I detection chemistry.
No, the primer pairs have been specifically designed to detect mRNA transcripts and their corresponding cDNA. However, primer pairs with a ranking of 4 may also co-amplify contaminating genomic DNA.
Sequence information will be provided on the Technical Data Sheets that accompany each primer pair shipment. 
No, only one forward and one reverse primer, dry, in individual tubes is included.
Possibly, but the primer pairs were designed to be species specific.
Both transcriptomes and genomes.
Depending on the species, it was either the amplicons and / or the primers.
The best approach would be to mention the species, gene, gene ID, primer pair (1, 2, or 3). Let us know when you publish, and we will send you a gift.