Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

578703

Sigma-Aldrich

Dichloro(3-methyl-2-butenylidene)bis(tricyclopentylphosphine)ruthenium(II)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C35H62Cl2P2Ru
CAS Number:
Molecular Weight:
716.79
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

solid

mp

94-96 °C (lit.)

storage temp.

2-8°C

SMILES string

Cl[Ru]Cl.[CH][CH][C](C)C.C1CCC(C1)P(C2CCCC2)C3CCCC3.C4CCC(C4)P(C5CCCC5)C6CCCC6

InChI

1S/2C15H27P.C5H8.2ClH.Ru/c2*1-2-8-13(7-1)16(14-9-3-4-10-14)15-11-5-6-12-15;1-4-5(2)3;;;/h2*13-15H,1-12H2;1,4H,2-3H3;2*1H;/q;;;;;+2/p-2

InChI key

UCDVULRIJNJVEV-UHFFFAOYSA-L

Application

Metathesis: Ruthenium-Based Metathesis Catalysts

  • Reactant for synthesis of ruthenium olefin metathesis catalysts

  • Catalyst for ring-opening metathesis polymerization
Kinetic application profile similar to that of Grubbs 1st Generation Catalyst.

Other Notes

NO BULK ORDERS

Legal Information

US Patent No. 5,969,170 and foreign equivalents apply. Sale of this product conveys to the buyer a limited-use research license. For questions, please contact us at aldrich@sial.com

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Furstner, A.
Angewandte Chemie (International Edition in English), 3012-3012 (2000)
Vougioukalakis, G. C.; Grubbs, R. H.
Chemical Reviews, 1746-1746 (2010)
Grubbs, R. H.
Tetrahedron, 7117-7117 (2004)
Trnka, T. M.; Grubbs, R. H.
Accounts of Chemical Research, 18-18 (2001)
Schuster, M.; Blechert, S.
Angewandte Chemie (International Edition in English), 2036-2036 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service