Skip to Content
Merck
All Photos(1)

Key Documents

704245

Sigma-Aldrich

Lanthanum strontium manganite

composite cathode powder, LSM-20/YSZ, ≥99%

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
26111700

Assay

≥99%

form

powder

composition

(Y2O3)0.08(ZrO2)0.92, 50 wt. %
La0.8Sr0.2MnO3, 50 wt. %

application(s)

battery manufacturing

Looking for similar products? Visit Product Comparison Guide

Application

LSM mixed with yttria-stabilized zirconia (YSZ) can be used in the cathodes for solid oxide fuel cells (SOFCs).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advances in Materials for Solid Oxide Fuel Cells
Gorte RJ
Material Matters, 5(4) null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service