803936
Poly [silicone-alt-PEG] dimethacrylate
average Mn 3,000, contains MEHQ as inhibitor
Synonym(s):
PEG-PDMS block copolymer, PEG-PDMS methacrylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C7H11O2)2 C8H22O2Si3 [C14H34O6Si3]n
UNSPSC Code:
12162002
Recommended Products
mol wt
average Mn 3,000
contains
MEHQ as inhibitor
storage temp.
−20°C
Application
This macromonomer can be used in many applications, including applications where gas permeability is desired, such as ophthalmic applications.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service