900707
Graphene quantum dots
cyan luminescent, 1 mg/mL in H2O
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
description
topographic height: 1-2.0 nm
form
suspension
concentration
1 mg/mL in H2O
color
colorless
diameter
<5 nm
fluorescence
λem 475-495 nm FWHM 70 nm, quantum yield >17%
InChI
1S/C
InChI key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Graphene quantum dots (GQDs), sheets of few-layered graphene and lateral dimensions smaller than 100 nm, possess strong quantum confinement and edge effects. Thus, they possess unique physical properties such as strong photoluminescence, which can be tailored for specific applications by controlling their size, shape, defects, and functionality. In contrast to classic QDs, such as metal or silicon quantum dots, GQDs are biocompatible, photo-stable, and inherit superior thermal, electrical, and mechanical properties from the graphene. These features can greatly contribute to various state-of-the-art applications:
- Optical brighteners.
- Taggants for security application.
- Bio-imaging markers.
- Fluorescent polymers.
- Antibacterial, antibiofouling, disinfection systems.
- Heavy metals, humidity and pressure sensors.
- Batteries.
- Flash memory devices.
- Photovoltaic devices.
- Light-emitting diodes.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Rostislav Bukasov et al.
Journal of fluorescence, 30(6), 1477-1482 (2020-08-29)
Surface enhanced fluorescence (SEF) is observed with very high contrast (100-200) from single E. coli bacteria cells labeled with Carbon nanodots (CDs), on aluminum foil and aluminum film. Likely, it is the first application of organic CDs in SEF. SEF
Y Ito et al.
Clinica chimica acta; international journal of clinical chemistry, 194(2-3), 131-144 (1990-12-24)
Serum concentrations of alpha-carotene (AC), beta-carotene (BC), lycopene (LY), beta-cryptoxanthin (CR), zeaxanthin (including lutein. ZX), canthaxanthin (CX), retinol (RE), and alpha-tocopherol (TO) in healthy persons (618 males and 1,196 females) aged 7-86 years were determined by HPLC. Serum concentrations of
Jianhua Shen et al.
Chemical communications (Cambridge, England), 48(31), 3686-3699 (2012-03-14)
Similar to the popular older cousins, luminescent carbon dots (C-dots), graphene quantum dots or graphene quantum discs (GQDs) have generated enormous excitement because of their superiority in chemical inertness, biocompatibility and low toxicity. Besides, GQDs, consisting of a single atomic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service