Skip to Content
Merck
All Photos(2)

Key Documents

901937

Sigma-Aldrich

NORIT® SX Plus CAT

Synonym(s):

Activated carbon, Activated charcoal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C
CAS Number:
UNSPSC Code:
23201100
NACRES:
NA.23

description

Methylene blue adsorption 20g/100 g

form

powder

surface area

1000 m2/g , Total

bulk density

200‑300 kg/m3

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Application

  • Activated carbons are internally porous microcrystalline, non-graphitic forms of carbon. The activated carbons possess large surface area (~1000 m2/g) and pore volumes which make them useful for applications in energy storage devices, catalysis, and for removal of impurities from gases and liquids.
  • NORIT® SX Plus CAT is a powdered activated carbon designed for use as carrier of precious metals in liquid phase pharmaceutical and fine chemical synthesis. Its unique raw material enhances the metal dispersion over its surface area which reduces the need of metal to achieve the same catalytic activity.
  • The high porosity of NORIT® SX Plus CAT results in a high adsorption capacity for a large range of impurities originating from both the reactants and by-products. Adsorption of impurities improves the purity of the end product and overcomes the issue of catalyst poisoning and leads to longer catalyst life. The NORIT® SX Plus CAT possesses narrow particles size distribution which leads to enhancement in filtration process and reduces the processing time.

Legal Information

Product of Cabot Corporation.
NORIT® is a registered trademark of Cabot Corporation.
Norit is a registered trademark of Norit N.V.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Water and Wastewater Treatment: Historical Perspective of Activated Carbon Adsorption and its Integration with Biological Processes.
Aktas O, et al.
Wiley-VCH (2011)
Energy storage applications of activated carbons: supercapacitors and hydrogen storage.
Sevilla M and Mokaya R
Energy & Environmental Science, 7, 1250-1280 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service