1.19257
Chlorine Powder Packs for photometers of other manufacturers for 10-ml samples
(total chlorine) Method: photometric, DPD 0 - 2.00 mg/l Cl₂ Spectroquant®
About This Item
Recommended Products
Quality Level
specific analyte(s)
chlorine
technique(s)
photometry: suitable
pH
6.3 (20 °C, 10 g/L in H2O)
storage temp.
15-25°C
Analysis Note
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Preparation of a standard solution for total chlorine.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service