KK1007
Roche
KAPA dNTP Kit (100 mM each)
Synonym(s):
DNPT, KAPA, PCR
Sign Into View Organizational & Contract Pricing
Select a Size
All Photos(1)
Select a Size
Change View
About This Item
UNSPSC Code:
41106300
NACRES:
NA.55
shelf life
≤18 mo.
packaging
pkg of 4 x 250 μL
manufacturer/tradename
Roche
storage temp.
−20°C
General description
KAPA dNTP Kit offers premium, high-quality, molecular grade dinucleotide triphosphates (dNTPs) used typically for polymerase chain reaction (PCR).
Application
KAPA dNTPs are suitable for use in any molecular biology application requiring high-quality dNTPs, such as:
- Standard polymerase chain reaction (PCR)
- High fidelity PCR
- Quantitative PCR
- End-repair or A-tailing
- cDNA synthesis
- DNA sequencing
- DNA labeling
- Microarrays
Features and Benefits
- KAPA dNTPs are available as individual dNTPs or as a ready-to-use dNTP Mix.
- KAPA dNTP Mix is supplied with KAPA Taq, KAPA2G Fast, KAPA2G Robust, KAPA LongRange and KAPA HiFi PCR Kits.
- KAPA dNTPs are subjected to strict quality control to ensure high performance, particularly with B-family polymerases that are sensitive to poisoning by uracil.
- Compatible with any commercially available DNA polymerases.
Quality
KAPA dNTPs are >99% pure by HPLC and are free of DNase, RNase, protease, phosphatase, and nicking activity. All kit components are subjected to stringent functional quality control, and meet strict requirements with respect to DNA contamination.
Preparation Note
Always ensure that the product has been fully thawed and mixed before use. KAPA dNTPs may be stored at 4 °C for short-term use (up to 1 month). Return to -20 °C for long-term storage. Provided that the sample has been handled carefully and not contaminated, the samples are not expected to be compromised if left (unintentionally) at room temperature for a short period of time (up to 3 days). Long-term storage at room temperature and 4 °C is not recommended. Please note that storing the reagents at temperatures above -20 °C makes them more prone to degradation when contaminated during use, and therefore storage at such temperatures is at the user′s own risk.
Other Notes
For Research Use Only. Not for use in diagnostic procedures.
Kit Components Only
Product No.
Description
- dATP 100 mM
- dCTP 100 mM
- dGTP 100 mM
- dTTP 100 mM
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service