Skip to Content
Merck
All Photos(1)

Key Documents

PHR8797

Lacosamide Related Compound D

new

certified reference material, pharmaceutical secondary standard

Synonym(s):

2-Amino-N-benzyl-3-methoxypropanamide oxalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N2O2 · C2H2O4
CAS Number:
Molecular Weight:
298.29
UNSPSC Code:
12352200

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to EP (Catalog No. Y0002065)
traceable to USP (Catalog No. 1357147)

API family

lacosamide

CofA

current certificate can be downloaded

packaging

pkg of 50 mg

application(s)

pharmaceutical

storage temp.

-10 to -25°C

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service