PHR8852
Clorazepate Dipotassium

certified reference material, pharmaceutical secondary standard
About This Item
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to USP (Reference Standard 1140509)
API family
clorazepate
CofA
current certificate can be downloaded
packaging
pkg of 250 mg
application(s)
pharmaceutical
storage temp.
-10 to -25°C
General description
Analysis Note
Other Notes
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Repr. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service