Skip to Content
Merck
All Photos(3)

Key Documents

NA0500

Sigma-Aldrich

GenElute HP Plasmid Megaprep Kit

sufficient for 5 preparations

Synonym(s):

Gen Elute, GenElute HP Plasmid Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.52

Pricing and availability is not currently available.

usage

sufficient for 5 preparations

greener alternative product characteristics

Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

General description

The GenElute HP Plasmid Megaprep Kit offers a simple, rapid, and cost-effective method for isolating plasmid DNA from recombinant E. coli cultures. This high performance kit features vacuum filter units for rapid lysate clearing and for DNA-silica binding. Up to 5 mg of high copy plasmid DNA can be recovered from 200 mL-1.0 L LB medium or 200-600 mL TB medium of E. coli culture in less than 1.5 hours. The actual yield depends on the strain, the plasmid, and the culture medium used.

An overnight recombinant E. coli culture is harvested with centrifugation and subjected to a modified alkaline-SDS lysis procedure followed by adsorption of the DNA onto a silica membrane in the presence of high salts. Contaminants are removed by two wash steps. Finally, the bound DNA is then eluted in Elution Solution (Tris-HCl) or water.
The GenElute HP Plasmid Megaprep Kit offers a simple, rapid, and cost-effective method for isolating plasmid DNA from recombinant E. coli cultures. This high performance kit features vacuum filter units for rapid lysate clearing and for DNA-silica binding. Up to 5 mg of high copy plasmid DNA can be recovered from 200 mL-1.0 L LB medium or 200-600 mL TB medium of E. coli culture in less than 1.5 hours. The actual yield depends on the strain, the plasmid, and the culture medium used.

An overnight recombinant E. coli culture is harvested with centrifugation and subjected to a modified alkaline-SDS lysis procedure followed by adsorption of the DNA onto a silica membrane in the presence of high salts. Contaminants are removed by two wash steps. Finally, the bound DNA is then eluted in Elution Solution (Tris-HCl) or water.

The DNA is ready for immediate use in applications such as restriction digest, cloning, sequencing, PCR, and transfection.

Application

GenElute HP Plasmid Megaprep Kit has been used to purify plasmids.[1]
The recovered plasmid DNA is predominately in its supercoiled form. Genomic DNA and RNA are below detectable levels by ethidium bromide stained agarose gel electrophoresis. The DNA is ready for immediate use in downstream applications such as restriction digestion, ligation, sequencing, PCR, transformation, and transfection.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of phenol and chloroform to perform DNA extractions.

Features and Benefits

  • From harvested bacterial culture to pure plasmid DNA in 90 minutes or less
  • Up to 5 mg of high-copy plasmid DNA
  • Efficient vacuum format
  • No phenol/chloroform extraction of alcohol precipitation required

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1B - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Lungs

Storage Class Code

3 - Flammable liquids

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Effect of acyl chain length on transfection efficiency and toxicity of polyethylenimine.
Aravindan L
International Journal of Pharmaceutics, 378(1-2), 201-210 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service