Key Documents
Z284777
Sigma-Aldrich
Screw-caps for Aldrich® fritted filter funnel
thread size 38-430
About This Item
UNSPSC Code:
41121800
NACRES:
NB.24
packaging
pkg of 10 ea
thread size
38-430
General description
Black plastic with PTFE liner.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)