Z567329
Wilmad® quartz (CFQ) EPR tubes
O.D. 2 mm, L 100 mm
Synonym(s):
Wilmad nmr tubes, nmr sample tubes, nmr tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
quartz tube
packaging
pack of 5 ea
manufacturer/tradename
Wilmad 712-SQ-100M
L
100 mm
O.D.
2 mm
tube diam.
2 mm
wall thickness
0.5 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Clear Fused-Quartz (CFQ), is a naturally occurring material. It has a high dielectric constant and low levels of paramagnetic impurities. This NMR tube is suitable for use in routine testing in EPR spectropscopy, excluding UV irradiation and samples with low sensitivity.
Legal Information
Wilmad is a registered trademark of SP Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service