Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S7905-M

Sigma-Aldrich

Amplifluor® UniPrimer II Fluorescein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.84

manufacturer/tradename

Chemicon®

Quality Level

100
300

technique(s)

PCR: suitable

application(s)

genomic analysis

detection method

fluorometric

General description

The Amplifluor® UniPrimer-fluorescein is a hairpin primer labeled with a fluorescent energy transfer pair (fluorescein and non-fluorescent quencher) and a 3′ single-stranded oligonucleotide tail and is provided at a concentration of 5.0 μM.

Fluorescence (Linear:Hairpin): 39. Excitation = 495nm; Emission = 516nm.

Application

Research Category
All

Physical form

5.0μM (2.5 mL)

Storage and Stability

Minimize exposure to light as this material is light sensitive. Make sterile aliquots of bulk solution. Store long term at -65°C to -75°C. Packaged product for end user may be stored in the dark at -15°C to -25°C.

Recommended diluent: autoclaved, reagent grade, deionized water only.

Legal Information

Amplifluor is a registered trademark of Merck KGaA, Darmstadt, Germany
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service