Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10736619001

Roche

Ascorbate Oxidase Spatula

from Cucurbita species

Synonym(s):

ascorbate oxidase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204

biological source

plant (Cucurbita species)

Quality Level

form

solid

specific activity

17 units

packaging

pkg of 25 ea

manufacturer/tradename

Roche

storage condition

protect from light

concentration

≥0.1-1.0 % (w/w)

technique(s)

activity assay: suitable

color

light gray

optimum pH

~6.0

pH range

5.6-7.0

solubility

water: partially soluble

suitability

suitable for HPLC

NCBI accession no.

UniProt accession no.

application(s)

food and beverages
life science and biopharma

storage temp.

2-8°C

Related Categories

General description

L-ascorbate:oxygen oxidoreductase

Specificity

Specific activity: One spatula contains approximately 17 U ascorbate oxidase at +25°C with L-ascorbate as the substrate.

Application

The spatula serves to remove ascorbic acid from aqueous solutions in which the presence of ascorbic acid (ascorbate, vitamin C) can interfere with the investigation of the sample material, for example, the detection of blood (erythrocytes) in urine or the determination of oxalic acid in urine during research studies.
The ascorbic acid to be removed should not exceed a concentration of 10 μg/ml. For concentrations in the range of 50 to 100 μg/ml, stir for 15 minutes. For higher concentrations, the solution should be appropriately diluted.

Physical form

Spatulas

Storage and Stability

Store at 2 to 8 °C. (Store dry!)

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service