Skip to Content
Merck
All Photos(1)

Key Documents

72963

Sigma-Aldrich

12-(7-Nitrobenzofurazan-4-ylamino)dodecanoic acid

suitable for fluorescence

Synonym(s):

12-(7-Nitro-2,1,3-benzoxadiazol-4-ylamino)dodecanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H26N4O5
CAS Number:
Molecular Weight:
378.42
Beilstein:
7392043
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98.0% (TLC)

form

powder

mp

79-81 °C (lit.)

solubility

DMF: soluble
methanol: soluble

fluorescence

λex 466 nm; λem 530 nm in methanol

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

OC(=O)CCCCCCCCCCCNc1ccc([N+]([O-])=O)c2nonc12

InChI

1S/C18H26N4O5/c23-16(24)10-8-6-4-2-1-3-5-7-9-13-19-14-11-12-15(22(25)26)18-17(14)20-27-21-18/h11-12,19H,1-10,13H2,(H,23,24)

InChI key

YPOHVKMXKUSZRL-UHFFFAOYSA-N

Other Notes

For probing the ligand binding sites of fatty acid and sterol carrier proteins

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Schroeder et al.
Biochemistry, 34(37), 11919-11927 (1995-09-19)
Direct effects of ethanol on the interaction of cytosolic lipid transfer proteins with ligands are not known. In this study, recombinant liver fatty acid binding protein (L-FABP) and sterol carrier protein-2 (SCP-2) were used in conjunction with a series of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service