Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PE117 human cell line

IHW Number: 9028, 88052028

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human blood

ethnicity

Native American

gender

male

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblastoid

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

Looking for similar products? Visit Product Comparison Guide

General description

Consanguineous: No

Cell Line Description

This cell line is an Epstein-Barr Virus (EBV) transformed lymphoblastoid line and is part of the Human Leukocyte Antigen (HLA) Typed Collection maintained by the European Collection of Cell Cultures (ECACC). HLA-Typing Data is available for this cell line.

DNA Profile

A: 2402
Cw: 304
B: 4001 4002
HLA DRA: 101
HLA DRB1: 404
HLA DRB4: 101
HLA DQA1: 3
HLA DQB1: 302
HLA DPA1: 1
HLA DPB1: 401

Serological Profile

HLA A: 24
HLA A ief: 24.1
HLA C: 10
HLA B: 60 61
HLA B ief: 60 61
Bw4/bw6: 6
HLA Dw: 14
HLA DR: 4
HLA DR53: 53
HLA DQ: 8
HLA DP: 4

Culture Medium

RPMI 1640 (R0883) + 2mM Glutamine (G7513) + 20% Foetal Bovine Serum (FBS) (F2442). Once a growing culture is established the FBS concentration can be reduced to 10%.

Subculture Routine

Maintain cultures between 3x105 - 2x106 cells/ml, 5% CO2, 37 °C. Upon resuscitation remove cryoprotectant - centrifuge at 300 x g for 5 minutes and re-suspend the cell pellet in fresh culture medium.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has restrictions and can only be supplied to academic and not-for-profit organisations. Nucleic acids can be supplied to all researchers including those from commercial organisations.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service