Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JY human cell line

IHW Number: 9287, 94022533

Synonym(s):

JY-S

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

biological source

human blood

ethnicity

Amish

gender

male

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblastoid

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

Looking for similar products? Visit Product Comparison Guide

General description

Consanguineous: No
Homozygous: No

Cell Line Description

This cell line is an Epstein-Barr Virus (EBV) transformed lymphoblastoid line and is part of the Human Leukocyte Antigen (HLA) Typed Collection maintained by the European Collection of Cell Cultures (ECACC). HLA-Typing Data is available for this cell line.

DNA Profile

A: 201
Cw: 702
B: 702
HLA DQA1: 3011
HLA DPB1: 201

Serological Profile

HLA A: 2
HLA C: 7
HLA B: 7
Bw4/bw6: 6
HLA DR: 4 13
HLA DR52: 52
HLA DR53: 53
HLA DQ: 1 3

Culture Medium

RPMI 1640 (R0883) + 2mM Glutamine (G7513) + 20% Foetal Bovine Serum (FBS) (F2442). Once a growing culture is established the FBS concentration can be reduced to 10%.

Subculture Routine

When resuscitating from frozen, cells should be seeded at 0.5 x 106 cells/ml in the culture medium. The culture should establish log phase growth after 3-5 days. When growing well dilute every 1-3 days to 3 x 105 cells/ml. Maintain cultures between 3-9 x 105 cells/ml for maximum growth; 5% CO2; 37°C. Cells form floating aggregates of varying size.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has restrictions and can only be supplied to academic and not-for-profit organisations. Nucleic acids can be supplied to all researchers including those from commercial organisations.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service