Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0572

Sigma-Aldrich

TIGIT, FC Fusion

recombinant, expressed in HEK 293 cells

Synonym(s):

T-cell immunoreceptor with Ig and ITIM domains, V-set and immunoglobulin domain-containing protein 9, V-set and transmembrane domain-containing protein 3, VSIG9, VSTM3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

39.7 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TIGIT(201633)

General description

Human T-cell immunoreceptor with Ig and ITIM domains (TIGIT), also known as V-set and immunoglobulin domain-containing protein 9, VSIG9, V-set and transmembrane domain-containing protein 3, and VSTM3, GenBank Accession No. NM_173799, a.a. 22-141 fused to Fc region of human IgG,expressed in a HEK293 cell expression system. MW = 39.7 kDa. This protein runs at a higher MW by SDS-PAGE due to glycosylation.

Physical form

Aqueous solution

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service