Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4739

Sigma-Aldrich

PLGF-1 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

PGF, PIGF, PLGF 1, Placenta Growth Factor-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

~29.7 kDa

packaging

pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PGF(5228)

General description

PLGF (placental growth factor), originally identified in human placenta, is a member of the VEGF (vascular endothelial growth factor) family. This protein is predominantly expressed in placenta, and also shows expression in heart, thyroid, lung and skeletal muscle. It is alternatively spliced to produce three isoforms in humans namely, PLGF1, PLGF2 and PLGF3.
Recombinant mouse PLGF is a disulfide-linked homodimer containing two 142 amino acid polypeptide chains.

Biochem/physiol Actions

PLGF (placental growth factor) is involved in angiogenesis, but does not play an essential role in angiogenesis during reproduction and development. However, inactivation of this gene in pathological conditions such as, ischemia, inflammation and cancer, results in impaired angiogenesis. Preclinical studies show that this protein might be a therapeutic target for treatment of angiogenesis and inflammation related disorders. In acute coronary syndrome (ACS) patients, increased levels of PLGF are linked with obstructive sleep apnea (OSA) and short-term adverse outcomes.

Physical form

Sterile filtered and lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute the protein in sterile 20 mM acetic acid to a concentration of 0.1-1.0 ug/ul. The solution can then be diluted into other aqueous buffer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impact of Obstructive Sleep Apnea on the Levels of Placental Growth Factor (PlGF) and Their Value for Predicting Short-Term Adverse Outcomes in Patients with Acute Coronary Syndrome.
Barcelo A, et al.
PLoS ONE, 11(3) (2016)
Placental growth factor and its receptor, vascular endothelial growth factor receptor-1: novel targets for stimulation of ischemic tissue revascularization and inhibition of angiogenic and inflammatory disorders.
Autiero M, et al.
Journal of Thrombosis and Haemostasis, 1(7), 1356-1370 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service