Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z744113

Freezer box divider

to hold, 64 tube(s), pk of 12

Synonym(s):

freezer container, freezer storage, storage container

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122804

feature

to hold 64 tube(s)

packaging

pack of 12
pk of 12

manufacturer/tradename

Diversified Biotech FBCD-1000

Looking for similar products? Visit Product Comparison Guide

General description

Freezer box divider, 64 hole 12/pk

Application

Cryogenics

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service