Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51080AST

Supelco

Astec® Diol (5 μm) HPLC Columns

L × I.D. 25 cm × 4.6 mm, HPLC Column

Synonym(s):

Diol Stationary Phase Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

product name

Astec® Diol HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm

material

stainless steel column

Agency

suitable for USP L20

product line

Astec®

packaging

pkg of 1 ea

manufacturer/tradename

Astec®

parameter

80-400 bar pressure (1200-5800 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

surface area

320 m2/g

matrix

fully porous particle
silica gel high purity, spherical particle platform

matrix active group

diol phase

particle size

5 μm

pore size

110 Å pore diameter

operating pH

3.5-7

separation technique

hydrophilic interaction (HILIC)
normal phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service