Skip to Content
Merck

ABD137

Anti-ESRRB Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Steroid hormone receptor ERR2, ERR beta-2, Estrogen receptor-like 2, Estrogen-related receptor beta, ERR-beta, Nuclear receptor subfamily 3 group B member 2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ESRRB(2103)

General description

Steroid hormone receptor ERR2, also known as ERR beta-2 or Estrogen receptor-like 2 or Estrogen-related receptor beta (ERR-beta), and encoded by the gene name ESRRB or ERRB2 or ESRL2, was initially identified in mouse embryonic carcinoma stem cells and was found to be expressed exclusively in trophoblast progenitor cells between days 6.5 and 7.5 post coitum. Recent studies have shown that ESRRB is part of an POU5F1/Oct4-centered protein interaction network in embryonic stem (ES) cells. It is thought to interact with other ES transcription factors such as NANOG within this network, activating POU5F1 transcription and thereby sustaining self-renewal and pluripotency in ES cells. Mutations in ESRRB have been associated with autosomal recessive deafness which is a form of non-syndromic deafness characterized by non-progressive, prelingual hearing loss.
~59 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Immunogen

Epitope: N-terminus
KLH-conjugated synthetic peptide corresponding to the N-terminus of human ESRRB.

Application

Anti-ESRRB Antibody detects level of ESRRB & has been published & validated for use in ESRRB.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Biochem/physiol Actions

Uniprot describes 2 isoforms produced by alternative splicing at ~56 kDa and 1 isoform at ~48 kDa.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in Human heart tissue lysate with and without blocking peptide.

Western Blotting Analysis: 1 μg/mL of this antibody detected ESRRB in Human heart tissue lysate with and without blocking peptide.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Explore all of our products under Anti-ESRRB Antibody

or

Try our Product Selector Tool to narrow your options


wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library