Skip to Content
Merck
All Photos(1)

Key Documents

763713

Sigma-Aldrich

Graphene oxide

sheets

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CxOyHz
UNSPSC Code:
12352103
NACRES:
NA.23

form

powder or flakes

SMILES string

O=C(O)C1C2=C3C4=C5C6=C7C8=C9C%10=C%11C(C%12=C%13C%10=C%14C8=C%15C6=C%16C4=C%17C2=CC(C(O)=O)C%18=C%17C%19=C%16C%20=C%15C%21=C%14C%22=C%13C(C%23=C%24C%22=C%25C%21=C%26C%20=C%27C%19=C%28C%18=CC(C(O)=O)C%29=C%28C%30=C%27C%31=C%26C%32=C%25C%33=C%24C(C%34=C%35C

InChI

1S/C140H42O20/c141-131(142)26-13-23-15-44-62(140(159)160)45-16-24-14-40-31(132(143)144)5-1-29-41-20-48(135(149)150)56-33-7-3-28-27-2-6-32-55-37(133(145)146)11-9-35-60(138(155)156)42-17-25-18-43-61(139(157)158)36-10-12-38(134(147)148)58-46-21-50(137(153)154)59-47-22-49(136(151)152)57-34-8-4-30-39(19-26)51(23)78-72(44)88-75(45)80-52(24)79(54(29)40)95-71(41)83(56)101-93-69(33)64(28)91-90-63(27)68(32)92-86(66(35)55)73(42)81-53(25)82-74(43)87(67(36)58)96-76(46)85(59)103-97-77(47)84(57)102-94-70(34)65(30)89(78)105-104(88)115-98(80)111(95)116(101)126-122-110(93)107(91)120-119-106(90)108(92)99(81)114-100(82)112(96)118(103)128(124(114)119)123-113(97)117(102)127(130(122)129(120)123)121(109(94)105)125(115)126/h2,5,7-10,12-22,26,38,48-50H,1,3-4,6,11H2,(H,141,142)(H,143,144)(H,145,146)(H,147,148)(H,149,150)(H,151,152)(H,153,154)(H,155,156)(H,157,158)(H,159,160)

InChI key

VTWITIAIMADGRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Graphene oxide (GO) is the solution processable precursor for bulk production of graphene based materials and devices. It is produced by the chemical exfoilation of graphite. GO is amphiphillic in nature, it also forms highly stable emulsions of organic solvents. Chemical modification of GO helps in improving the biocompatibility and solubility of graphene.

Application

Dielectric Materials for Field Effect Transistors (FET); Non-silicon-based materials for integrated circuits (IC). Some proposed applications of GO are:
  • water soluble/insoluble, targeted/untargeted drug and gene delivery applications
  • tissue engineering
  • electrochemical biosensors
  • optical biosensors
  • graphene oxide Langmuir Blodgett films as hole injection layers for organic light emitting diode.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Maria P Georgopoulou et al.
Colloids and surfaces. B, Biointerfaces, 189, 110841-110841 (2020-02-15)
This study examines the effect of graphene oxide (GO) nanoparticles (NPs) on the transport (individual species) and cotransport (simultaneous transport) of three biocolloids (Escherichia (E.) coli, Enterococcus (E.) faecalis and Staphylococcus (S.) aureus) in water saturated porous media. Flowthrough experiments
Tunable assembly of GO surfactant sheets: wrinkles, overlaps and impacts on thin film properties
Cote LJ, et al
Soft Matter, 6(24), 6096-6101 null
Jaemyung Kim et al.
Accounts of chemical research, 45(8), 1356-1364 (2012-06-06)
Graphite oxide sheets, now called graphene oxide (GO), can be made from chemical exfoliation of graphite by reactions that have been known for 150 years. Because GO is a promising solution-processable precursor for the bulk production of graphene, interest in
Osamah Al Rugaie et al.
Microorganisms, 9(1) (2021-01-21)
Gold nanoparticles (GNPs) and graphene oxide flakes (GOFs) exerted significantly (p < 0.0001) supportive roles on the phagocytosis bioactivity of the immune cells of phagocytic nature against the Gram-positive and Gram-negative human pathogenic bacteria Staphylococcus aureus and Escherichia coli. Under
Graphene in biotechnology
Material Matters, 8(1) null

Articles

3D printing is a type of additive manufacturing that can be used to rapidly fabricate components with highly customizable geometries.

Professor Rivnay (Northwestern University, USA) discusses using organic mixed conductors as an alternative to efficiently bridge the ionic world of biology with contemporary microelectronics.

Developed in the last several years, fluorescence quenching microscopy (FQM) has enabled rapid, inexpensive, and high-fidelity visualization of two-dimensional (2D) materials such as graphene-based sheets and MoS2.

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service