Skip to Content
Merck
All Photos(1)

Key Documents

I9531

Sigma-Aldrich

Imiloxan hydrochloride

>98% (HPLC), solid

Synonym(s):

2-[(2,3-dihydro-1,4-benzodioxin-2yl)methyl]-1-ethyl-1H-imidazole hydrochloride, RS 21361

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N2O2 · HCl
CAS Number:
Molecular Weight:
280.75
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

>98% (HPLC)

form

solid

color

white

solubility

H2O: soluble 30 mg/mL

SMILES string

Cl[H].CCn1ccnc1CC2COc3ccccc3O2

InChI

1S/C14H16N2O2.ClH/c1-2-16-8-7-15-14(16)9-11-10-17-12-5-3-4-6-13(12)18-11;/h3-8,11H,2,9-10H2,1H3;1H

InChI key

ANDJPJNFVJXEKX-UHFFFAOYSA-N

Gene Information

human ... ADRA2B(151)

Biochem/physiol Actions

Selective α2B-adrenoceptor antagonist.

Features and Benefits

This compound is featured on the α2-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fatih İlkaya et al.
Pharmacology, biochemistry, and behavior, 122, 1-6 (2014-03-04)
It has been shown that imipramine, a tricyclic antidepressant (TCA), is a potent analgesic agent. However, the effect of imipramine on visceral pain has not been extensively investigated. In the current study, our aim was to characterise the putative analgesic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service