798118
Poly(dimethylsiloxane), monoacrylamidopropyl terminated
Mn 1,200, contains MEHQ as inhibitor
Synonym(s):
Acrylamide silicone, PDMS acrylamide
Sign Into View Organizational & Contract Pricing
Select a Size
All Photos(1)
Select a Size
Change View
About This Item
Linear Formula:
C8H16NO2Si(C2H6OSi)nSiC6H15
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
form
liquid
mol wt
Mn 1,200
contains
MEHQ as inhibitor
density
0.954 g/mL at 25 °C
storage temp.
−20°C
Application
This macromonomer can be used in many applications including ophthalmic applications where gas permeability is desired. This methacrylamide can also be polymerized using controlled radical polymerization such as ATRP or RAFT.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service