72615
Acrylamide Kit I
for the determination of acrylamide acc. to the Grob method
About This Item
Recommended Products
quality
for the determination of acrylamide acc. to the Grob method
application(s)
cleaning products
cosmetics
food and beverages
personal care
storage temp.
2-8°C
Components
acrylamide-d3 standard solution (500 mg/L),
methacrylamide standard solution (500 mg/L),
butyramide standard solution (25 mg/L),
1-propanol, n-hexane, acetonitrile and oil.
All kit components are suitable for acrylamide determination according to Grob.
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service