Skip to Content
Merck
All Photos(1)

Key Documents

PHR9319

Supelco

(17alpha)-Hydroxy-13(17)a-homoprednisolone

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

((2R,3R,4aS,4bS,10aR,10bS,11S,12aS)-2,3,11-Trihydroxy-2-(hydroxymethyl)-10a,12a-dimethyl3,4,4a,5,6,10a,10b,11,12,12a-decahydrochrysene-1,8(2H,4bH)-dione)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O6
Molecular Weight:
376.44
UNSPSC Code:
41116107

grade

certified reference material
TraceCERT®

Quality Level

Agency

(Not traceable to any primary pharmacopeia standard)

API family

prednisolone, prednisone

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

application(s)

pharmaceutical

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Other Notes

Budesonide Impurity

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service