Merck
GC Analysis of L-Carvone Enantiomer in Scotch Spearmint Essential Oil on Astec® CHIRALDEX G-TA
GC Analysis of L-Carvone Enantiomer in Scotch Spearmint Essential Oil on Astec® CHIRALDEX™ G-TA suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX G-TA Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
CONDITIONS
column
Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 μm (73033AST)
oven
40 °C (1 min), 2 °C/min to 170 °C (15 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 25 cm/sec, set at 110 °C
injection
0.1 μL 100:1 split
liner
4 mm I.D., split type, cup design
sample
Scotch Spearmint Oil, neat
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC