Skip to Content
Merck
All Photos(1)

Key Documents

B3676

Millipore

B.T.B. lactose agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Bromo Thymol Blue

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

product line

BioChemika

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | utilization test: suitable
microbiological culture: suitable

final pH

8.6±0.2 (25 °C)

application(s)

bioburden testing
cleaning products
cosmetics
food and beverages
personal care
personal care

microbiology

suitability

nonselective and differential for Staphylococcus spp.

Application

Used for the detection and isolation of pathogenic Staphylococci.

Components

Ingredients (g/L):
Proteose peptone, 5.00
Beef extract, 3.00
Lactose, 10.00
Bromo Thymol Blue, 0.17
Agar, 15.00

Preparation Note

Suspend 33.17 g of B.T.B. Lactose Agar in 1000 ml of distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Chromogenic media enable the selective detection of S. aureus, which produce bluish-green colonies that are clearly differentiated from other species.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service