318477
Indium(III) hydroxide
99.99% trace metals basis
About This Item
Recommended Products
vapor pressure
<0.01 mmHg ( 25 °C)
Assay
99.99% trace metals basis
form
powder and chunks
reaction suitability
reagent type: catalyst
core: indium
impurities
≤200 ppm Trace metallic impurities analysis (ICP)
mp
150 °C (lit.)
solubility
H2O: insoluble(lit.)
density
4.45 g/mL at 20 °C (lit.)
SMILES string
O[In](O)O
InChI
1S/In.3H2O/h;3*1H2/q+3;;;/p-3
InChI key
IGUXCTSQIGAGSV-UHFFFAOYSA-K
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - STOT RE 1
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service