Skip to Content
Merck
All Photos(2)

Key Documents

389498

Sigma-Aldrich

4-Chloro-2-methoxyphenol

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(OCH3)OH
CAS Number:
Molecular Weight:
158.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

form

liquid

refractive index

n20/D 1.559 (lit.)

bp

241 °C (lit.)

mp

16-17 °C (lit.)

density

1.304 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

COc1cc(Cl)ccc1O

InChI

1S/C7H7ClO2/c1-10-7-4-5(8)2-3-6(7)9/h2-4,9H,1H3

InChI key

FVZQMMMRFNURSH-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Himadri Rajput et al.
Chemosphere, 218, 687-695 (2018-12-05)
In this study, novel Cesium (Cs) doped TiO2 nanotubes photoelectrode (Cs/TiO2NTs) were synthesized by simple electrochemical anodization method and characterized by several physicochemical techniques. In particular, the photocatalytic (PC), electrocatalytic (EC) and photoelectrocatalytic (PEC) activity of newly synthesized Cs/TiO2NTs electrodes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service