Skip to Content
Merck
All Photos(3)

Key Documents

527084

Sigma-Aldrich

Nafion perfluorinated resin solution

greener alternative

5 wt. % in mixture of lower aliphatic alcohols and water, contains 45% water

Synonym(s):

D-520

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
26111700
eCl@ss:
32110203
NACRES:
NA.23

eq. wt.

1,000

contains

45% water

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

5 wt. % in mixture of lower aliphatic alcohols and water

refractive index

n20/D 1.368

bp

91-104 °C

density

0.924 g/mL at 25 °C

greener alternative category

InChI

1S/C7HF13O5S.C2F4/c8-1(9)2(10)24-5(15,16)3(11,4(12,13)14)25-6(17,18)7(19,20)26(21,22)23;3-1(4)2(5)6/h(H,21,22,23);

InChI key

FOYUGSIADQEOEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Nafion perfluorinated resin solution is widely used as an electrolytic material. It can be used in the formation of a composite membrane with metal oxides. The membrane further facilitates the formation of chemically stable proton exchange membrane (PEM) fuel cells. It may be coated on mesoporous metallic films to improve the detection and selectivity of biomarkers. It may form a composite electrode material with graphene for glucose biosensors.
Support material for an integrated system which used solar energy to drive chemical reactions. Acts as a superacid catalyst for organic synthesis. When formed as metal composites, can be used as artificial muscle actuators, solid electrolytes for the analysis of electroactive species in organic media, or as electrodes in organic electrosynthesis.
Used to solubilize carbon nanotubes. Carbon nanotube solutions have been explored as amperometric biosensors.

Packaging

Packaged in glass bottles

Legal Information

Nafion is a trademark of The Chemours Company FC, LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 2 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph Wang et al.
Journal of the American Chemical Society, 125(9), 2408-2409 (2003-02-27)
The ability to solubilize single-wall and multiwall carbon nanotubes (CNT) in the presence of the perfluorinated polymer Nafion is described. Such use of Nafion as a solubilizing agent for CNT overcomes a major obstacle for creating CNT-based biosensing devices. Their
Glucose biosensor of ruthenium-doped TiO2 sensing electrode by co-sputtering system
Chou JC, et al.
Microelectronics reliability (2010)
F Pelayo García de Arquer et al.
Science (New York, N.Y.), 367(6478), 661-666 (2020-02-08)
Electrolysis offers an attractive route to upgrade greenhouse gases such as carbon dioxide (CO2) to valuable fuels and feedstocks; however, productivity is often limited by gas diffusion through a liquid electrolyte to the surface of the catalyst. Here, we present
An enzymatic glucose biosensor based on a glassy carbon electrode modified with manganese dioxide nanowires
Zhang L, et al.
Microchimica Acta, 54-56 (2013)
Nafion-coated mesoporous Pd film toward remarkably enhanced detection of lactic acid
Baba D, et al.
Royal Society of Chemistry Advances, 8(19), 10446-10449 (2018)

Articles

Polymer electrolyte membrane (PEM) fuel cells offer high efficiency and low-temperature operation, reducing oil use in light-duty vehicle transportation.

Proton exchange membrane (PEM) fuel cells operate at relatively low temperatures and are composed of two electrodes and a conductive elecrolyte.

Advances in electrochemical water conversion and understanding PEMFC degradation drive progress in hydrogen technologies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service