Skip to Content
Merck
All Photos(1)

Key Documents

566225

Sigma-Aldrich

7-[4-(Trifluoromethyl)coumarin]methacrylamide

98%

Synonym(s):

7-(Methacryloylamino)-4-(trifluoromethyl)coumarin, N-(2-Oxo-4-(trifluoromethyl)-2H-chromen-7-yl)methacrylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10F3NO3
CAS Number:
Molecular Weight:
297.23
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

mp

167-171 °C (lit.)

λmax

341 nm

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)Nc1ccc2c(OC(=O)C=C2C(F)(F)F)c1

InChI

1S/C14H10F3NO3/c1-7(2)13(20)18-8-3-4-9-10(14(15,16)17)6-12(19)21-11(9)5-8/h3-6H,1H2,2H3,(H,18,20)

InChI key

UDIRYDKCHVDUQV-UHFFFAOYSA-N

Application

Fluorescent monomer

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos Carbonell et al.
Nature communications, 11(1), 1244-1244 (2020-03-08)
Polymer brush patterns have a central role in established and emerging research disciplines, from microarrays and smart surfaces to tissue engineering. The properties of these patterned surfaces are dependent on monomer composition, polymer height, and brush distribution across the surface.
Zhaoyang Ding et al.
Polymers, 10(3) (2019-04-11)
Temperature is a significant parameter to regulate biological reactions and functions inside cells. Sensing the intracellular temperature with a competent method is necessary to understand life science. In this work, an energy-transfer polymeric thermometer was designed for temperature sensing. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service