Skip to Content
Merck
All Photos(2)

Key Documents

578789

Sigma-Aldrich

5,6-Epoxy-5,6-dihydro-[1,10]phenanthroline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8N2O
CAS Number:
Molecular Weight:
196.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

183 °C (dec.) (lit.)

functional group

ether

SMILES string

O1C2C1c3cccnc3-c4ncccc24

InChI

1S/C12H8N2O/c1-3-7-9(13-5-1)10-8(4-2-6-14-10)12-11(7)15-12/h1-6,11-12H

InChI key

GGQHROHZSIPVQE-UHFFFAOYSA-N

Application

5,6-Epoxy-5,6-dihydro-[1,10]phenanthroline may be used to synthesize N-(3-azidopropyl)-1,10-phenanthrolin-5-amine (az-phen), via reaction with 3-azidopropylamine which is then subjected to dehydration by treatment with sodium hydride. It can also be used to synthesize (1,10-phenanthrolin-5-yl)-1-thio-β-D-glucopyranoside by reacting with 1-thio-β-D-glucopyranose sodium salt in dry ethanol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thio-?-D-glucosides: Synthesis and Evaluation as Glycosidase Inhibitors and Activators
Samoshin.VA, et al.
International Journal of Carbohydrate Chemistry, 2014 (2014)
"A click chemistry based coordination polymer inside small heat shock protein"
Lucon J, et al
Chemical Communications (Cambridge, England), 46(02), 264- 266 (2010)
G M Lacourciere et al.
Biochemistry, 32(10), 2610-2616 (1993-03-16)
A recombinant baculovirus (vEHX) encoding rat hepatic microsomal epoxide hydrolase has been constructed. Infection of Spodoptera frugiperda (Sf9) cells with the recombinant virus results in the expression of the enzyme at a level estimated to be between 5% and 10%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service