697133
2-Amino-3-pyridinecarbonitrile
97%
Synonym(s):
2-Amino-3-cyanopyridine
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
C5H3NNH2CN
CAS Number:
Molecular Weight:
119.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
powder
mp
132-136 °C
functional group
nitrile
SMILES string
Nc1ncccc1C#N
InChI
1S/C6H5N3/c7-4-5-2-1-3-9-6(5)8/h1-3H,(H2,8,9)
InChI key
YYXDQRRDNPRJFL-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service