Skip to Content
Merck
All Photos(1)

Key Documents

Q1255

Sigma-Aldrich

Quinoline

reagent grade, 96%

Synonym(s):

1-Benzazine, 2,3-Benzopyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7N
CAS Number:
Molecular Weight:
129.16
Beilstein:
107477
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

vapor density

4.5 (vs air)

vapor pressure

0.07 mmHg ( 20 °C)

Assay

96%

autoignition temp.

896 °F

refractive index

n20/D 1.625 (lit.)

bp

113-114 °C/11 mmHg (lit.)
237 °C (lit.)

mp

−17-−13 °C (lit.)

density

1.093 g/mL at 25 °C (lit.)

SMILES string

c1ccc2ncccc2c1

InChI

1S/C9H7N/c1-2-6-9-8(4-1)5-3-7-10-9/h1-7H

InChI key

SMWDFEZZVXVKRB-UHFFFAOYSA-N

Gene Information

human ... CYP2D6(1565)

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margarita A Gonzalez et al.
Inorganic chemistry, 51(21), 11930-11940 (2012-10-24)
The unusual role of CO as a signaling molecule in several physiological pathways has spurred research in the area of synthesizing new CO-releasing molecules (CORMs) as exogenous CO donors. Auxiliary control on CO delivery can be achieved if CO can
Yoshiaki Yasutake et al.
FEBS letters, 588(1), 105-110 (2013-11-26)
The cytochrome P450 RauA from Rhodococcus erythropolis JCM 6824 catalyzes the hydroxylation of a nitrogen atom in the quinolone ring of aurachin, thereby conferring strong antibiotic activity on the aurachin alkaloid. Here, we report the crystal structure of RauA in
Rainer E Martin et al.
ChemMedChem, 8(4), 569-576 (2012-12-12)
Potency with potential: 2-Phenoxy-nicotinamides were identified as potent agonists at the GPBAR1 receptor, a target in the treatment of obesity, type 2 diabetes and metabolic syndrome. Extensive structure-activity relationship studies supported by homology modeling and docking resulted in the identification
R Musiol et al.
Current medicinal chemistry, 17(18), 1960-1973 (2010-04-10)
Although the assortment of antifungal drugs is broad, the most commonly used agents have major drawbacks. Toxicity, serious side effects or the emergence of drug resistance are amongst them. New drugs and drug candidates under clinical trials do not guarantee
Sergey Kapishnikov et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(28), 11188-11193 (2012-06-30)
Heme detoxification is a critical step in the life cycle of malaria-causing parasites, achieved by crystallization into physiologically insoluble hemozoin. The mode of nucleation has profound implications for understanding the mechanism of action of antimalarial drugs that inhibit hemozoin growth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service