181315-M
ARE Activator, BTZO-1
The ARE Activator, BTZO-1 modulates the biological activity of ARE. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Synonym(s):
ARE Activator, BTZO-1, 2-Pyridin-2-yl-4H-1,3-benzothiazin-4-one, Antioxidant Response Element Activator, Macrophage Migration Inhibitory Factor Antagonist II, MIF Antagonist II, Macrophage Migration Inhibitory Factor Antagonist II, MIF Antagonist II, 2-Pyridin-2-yl-4H-1,3-benzothiazin-4-one, Antioxidant Response Element Activator
About This Item
Assay
≥98% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
off-white
solubility
DMSO: 50 mg/mL
storage temp.
2-8°C
General description
Warning
Reconstitution
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service