Skip to Content
Merck
All Photos(1)

Key Documents

31146-M

Sigma-Aldrich

Boric acid

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, ≥99.8%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3BO3
CAS Number:
Molecular Weight:
61.83
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38120104
PubChem Substance ID:

grade

ACS reagent
puriss. p.a.

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor pressure

2.6 mmHg ( 20 °C)

Assay

≥99.8%

impurities

≤0.005% insoluble in methanol
≤0.05% with methanol-HCl non-volat. matter

pH

3.6-4.0 (20 °C, 4%)

mp

160 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤3 mg/kg
phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤5 mg/kg

cation traces

As: ≤0.00005%
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Cu: ≤0.0005%
Fe: ≤0.0001%
Mg: ≤0.0005%
Pb: ≤0.0005%
Zn: ≤0.0005%

SMILES string

OB(O)O

InChI

1S/BH3O3/c2-1(3)4/h2-4H

InChI key

KGBXLFKZBHKPEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Boric acid is a very weak acid, having pH in the range of 3.6-4.0. Coulometric method for the determination of boric acid has been reported.

Other Notes

Important notice
This product will be phased out towards early 2019. There is a replacement product available : 31146

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

L Charlet Bhami et al.
Journal of vector borne diseases, 52(2), 147-152 (2015-06-30)
The use of low concentrations of boric acid as a potential and effective control agent for the eggs and immature stages of Aedes aegypti L. and Aedes albopictus Skuse (Diptera: Culicidae) is found to be safe and effective as compared
L Romano et al.
The Journal of antimicrobial chemotherapy, 55(1), 110-114 (2004-12-03)
There is very little information, to date, on the antifungal activity of bergamot oil. In this study, we investigated the in vitro activity of three bergamot oils (natural essence, furocoumarin-free extract and distilled extract) against clinically relevant Candida species. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service