Skip to Content
Merck
All Photos(1)

Key Documents

2B300

Sigma-Aldrich

Sepharose 2B

60-200 μm bead diameter

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:

form

suspension

Quality Level

bead diameter

60-200 μm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Sepharose 2B is a well-proven cross-linked agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix. The matrix is not pre-activated and the user performs all steps in coupling.

Application

Sepharose 2B has been used for the preparation of virosomes and platelets.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Depletion of glycoprotein gp85 from virosomes made with Epstein-Barr virus proteins abolishes their ability to fuse with virus receptor-bearing cells.
Haddad RS and Hutt-Fletcher LM
Journal of Virology, 63, 4998-4998 (1989)
G Luzzatto et al.
Biochimica et biophysica acta, 1084(1), 78-86 (1991-06-19)
We have found that thrombin-induced activation of protein kinase C (PKC) in platelets, measured by phosphorylation of the 47 kDa protein, is synergistically enhanced by the amiloride analogue ethylisopropylamiloride (EIA), a specific inhibitor of Na+/H+ exchange. This EIA effect was
Molecular Cloning and Physical Mapping of Potato Virus S Complementary DNA.
Monis, J., and de Zoeten, G.A.
Phytopathology, 80, 446-450 (1990)
Balakrishnan Sivaraman et al.
Langmuir : the ACS journal of surfaces and colloids, 28(5), 2745-2752 (2011-12-24)
Recent studies have shown that platelets can adhere to adsorbed albumin (Alb) through a receptor-mediated mechanism, but only if the Alb undergoes more than a critical degree of adsorption-induced unfolding. The objectives of this research were to investigate whether Alb
A Sen et al.
Cell, 17(2), 347-356 (1979-06-01)
A low molecular weight (LMW) protein phosphokinase enzyme that binds to actin has been isolated from murine sarcoma virions; this kinase activity is not present in nontransforming murine leukemia viruses. Sephadex G-75 gel filtration and affinity chromatography on actin-Sepharose conjugates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service