Skip to Content
Merck
All Photos(1)

Key Documents

44026

Supelco

L-Phenylalanine hydrochloride solution

100 mM amino acid in 0.1 M HCl, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO2 · HCl
CAS Number:
Molecular Weight:
201.65
Beilstein:
8584529
MDL number:
UNSPSC Code:
85151701
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

form

liquid

analyte chemical class(es)

amino acids, peptides, proteins

concentration

100 mM amino acid in 0.1 M HCl

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

colorless

application(s)

food and beverages

format

single component solution

storage temp.

2-8°C

SMILES string

Cl.N[C@@H](Cc1ccccc1)C(O)=O

InChI

1S/C9H11NO2.ClH/c10-8(9(11)12)6-7-4-2-1-3-5-7;/h1-5,8H,6,10H2,(H,11,12);1H/t8-;/m0./s1

InChI key

ZAIZDXVMSSDZFA-QRPNPIFTSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service