Skip to Content
Merck
All Photos(1)

Key Documents

551392

Supelco

Triphenyl phosphate solution

NMR reference standard, 0.0485 M in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5O)3PO
Molecular Weight:
326.28
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24

grade

NMR reference standard
analytical standard

Quality Level

concentration

0.0485 M in chloroform-d (99.8 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

application(s)

environmental

format

single component solution

SMILES string

O=P(Oc1ccccc1)(Oc2ccccc2)Oc3ccccc3

InChI

1S/C18H15O4P/c19-23(20-16-10-4-1-5-11-16,21-17-12-6-2-7-13-17)22-18-14-8-3-9-15-18/h1-15H

InChI key

XZZNDPSIHUTMOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triphenyl phosphate is an important organophosphorus compoundand a colorless, crystalline, which can find applications in plasticizersand flame retardants industry. It is also used as a main component of lubricant oils and hydraulic fluids.

Features and Benefits

31P sensitivity

Quantity

5 mm O.D. tube contains 0.700 mL

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sullivan, BJ and Krieger, RG
Clinical Environmental Health and Toxic Exposures (2001)
Joint acute toxicity of tributyl phosphate and triphenyl phosphate to Daphnia magna
Lin K, et al.
Environmental Chemistry, 7, 309-309 (2009)
Studies on the thermal stabilization enhancement of ABS; synergistic effect of triphenyl phosphate nanocomposite, epoxy resin, and silane coupling agent mixtures
Kim J, et al.
Polymer Degradation and Stability, 79, 201-207 (2003)
Johanna Jyrkkärinne et al.
Journal of medicinal chemistry, 51(22), 7181-7192 (2008-11-06)
The human constitutive androstane receptor (CAR, NR1I3) is an important regulator of xenobiotic metabolism and other physiological processes. So far, only few CAR agonists are known and no explicit mechanism has been proposed for their action. Thus, we aimed to
M I Cervera et al.
Analytical and bioanalytical chemistry, 406(27), 6843-6855 (2014-05-16)
An atmospheric pressure chemical ionization source has been used to enhance the potential of gas chromatography coupled with quadrupole time-of-flight (QTOF) mass spectrometry (MS) for screening and quantification purposes in pesticide residue analysis. A screening method developed in our laboratory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service