Skip to Content
Merck
All Photos(1)

Key Documents

90733

Supelco

Quercetin 3-glucuronide

analytical standard

Synonym(s):

3,3′,4′,5,7-Pentahydroxyflavone 3-glucuronide, Mikwelianin, Miquelianin, Quercetin 3-O-β-D-glucuronide, Querciturone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18O13
CAS Number:
Molecular Weight:
478.36
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤15.0% water

application(s)

food and beverages

format

neat

SMILES string

O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C(O)=O)OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4

InChI

1S/C21H18O13/c22-7-4-10(25)12-11(5-7)32-17(6-1-2-8(23)9(24)3-6)18(13(12)26)33-21-16(29)14(27)15(28)19(34-21)20(30)31/h1-5,14-16,19,21-25,27-29H,(H,30,31)/t14-,15-,16+,19-,21+/m0/s1

InChI key

DUBCCGAQYVUYEU-ZUGPOPFOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Quercetin 3-glucuronide is a dietary flavanoid, that can be isolated from green beans.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

UV-induced biosynthesis of quercetin 3-O-?-D-glucuronide in dill cell cultures
Mohle B, et al.
Phytochemistry, 24, 465-467 (1985)
Absorption of quercetin-3-glucoside and quercetin-4?-glucoside in the rat small intestine: the role of lactase phlorizin hydrolase and the sodium-dependent glucose transporter
Day.JA, et al.
Biochemical Pharmacology, 65, 1199-1206 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service