Skip to Content
Merck
All Photos(1)

Key Documents

93544

Supelco

N-Hexanoylglycine

analytical standard

Synonym(s):

2-(Hexanoylamino)acetic acid, N-(1-Oxohexyl)glycine, N-Caproylglycine, Hexanoyl glycine, NSC 224460

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO3
CAS Number:
Molecular Weight:
173.21
Beilstein:
1773416
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

InChI

1S/C8H15NO3/c1-2-3-4-5-7(10)9-6-8(11)12/h2-6H2,1H3,(H,9,10)(H,11,12)

InChI key

UPCKIPHSXMXJOX-UHFFFAOYSA-N

Biochem/physiol Actions

Hexanoylglycine is an acyl glycine. Acyl glycines are normally minor metabolites of fatty acids. However, the excretion of certain acyl glycines is increased in several inborn errors of metabolism. In certain cases the measurement of these metabolites in body fluids can be used to diagnose disorders associated with mitochondrial fatty acid beta-oxidation. Acyl glycines are produced through the action of glycine N-acyltransferase (EC 2.3.1.13) which is an enzyme that catalyzes the chemical reaction:acyl-CoA + glycine ↔CoA + N-acylglycine. Hexanoylglycine is a fatty acid metabolite, it appears in the urine of patients with hereditary medium-chain acyl-CoA dehydrogenase (MCAD) deficiency.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Rinaldo et al.
Biomedical & environmental mass spectrometry, 18(7), 471-477 (1989-07-01)
We describe a gas chromatographic/mass spectrometric method for the accurate determination of n-hexanoylglycine, 3-phenylpropionylglycine and suberylglycine in urine for the diagnosis of hereditary medium-chain acyl-CoA dehydrogenase (MCAD) deficiency. These acylglycines had previously been detected in urine from patients with MCAD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service