Skip to Content
Merck
All Photos(1)

Documents

C9870

Sigma-Aldrich

Accellerase 1000

Synonym(s):

Cellulase from Trichoderma reesei

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

form

solid

concentration

≥1,000 unit/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

A commercial enzyme developed specifically to reduce complex lignocellulosic biomass into fermentable sugars.

Biochem/physiol Actions

Contains multiple glycolytic activities including exo-1,4-β− glucanase (cellobiohydrolase), endo-1,4-β-glucanase, hemicellulase and β-glucosidase.

Unit Definition

One unit will liberate 1.0 μmole of glucose from cellulose in one hr at pH 5.0 at 37 °C .

Legal Information

ACCELERASE is a trademark of Danisco US Inc.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service