Z637181
Mucasol™ universal detergent
volume 2 L
Synonym(s):
Mucasol Cleaner, Universal Detergent
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
BRAND 44003
technique(s)
protein expression: suitable
volume
2 L
20000 mL
Looking for similar products? Visit Product Comparison Guide
General description
Liquid, alkaline high-performance concentrate for immersion and ultrasonic baths.
The universal cleaning agent for labware and instruments made of glass, porclain, plastics, rubber and metal instruments. Items are cleaned within 10 to 30 minutes, without mechanical help. Mucasol replaces chromosulfuric acid, is free from chlorine and caustic alkalis, is non-toxic, non-aggressive and biodegradable.
Prolonged exposure to zinc and non-anodized aluminum surfaces should be avoided. Also suitable for cleaning by wiping.
The universal cleaning agent for labware and instruments made of glass, porclain, plastics, rubber and metal instruments. Items are cleaned within 10 to 30 minutes, without mechanical help. Mucasol replaces chromosulfuric acid, is free from chlorine and caustic alkalis, is non-toxic, non-aggressive and biodegradable.
Prolonged exposure to zinc and non-anodized aluminum surfaces should be avoided. Also suitable for cleaning by wiping.
Other Notes
BR44003 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Mucasol is a trademark of Merz GmbH & Co. KG
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service